2-Amino-5-(p-tolyl)thiazole 98%

Lieferant: Apollo Scientific

Synonyme: 5-(p-Tolyl)thiazol-2-amine

OR909075-1G OR909075-250MG
APOSOR909075-1GEA 290 EUR
APOSOR909075-1G APOSOR909075-250MG
2-Amino-5-(p-tolyl)thiazole 98%
2-Amino-5-(p-tolyl)thiazole
Formel: C₁₀H₁₀N₂S
Molecular Weight: 190,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00858269
CAS-Nummer: 73040-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR