N-t-Butyl pyrrole-2-carboxamide 98%

Lieferant: Apollo Scientific

Synonyme: N-(tert-Butyl)-1H-pyrrole-2-carboxamide

OR909327-1G OR909327-5G
APOSOR909327-1GEA 128 EUR
APOSOR909327-1G APOSOR909327-5G
N-t-Butyl pyrrole-2-carboxamide 98%
N-t-Butyl pyrrole-2-carboxamide
Formel: C₉H₁₄N₂O
Molecular Weight: 166.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16618952
CAS-Nummer: 1228957-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR