5-Chlorobenzo[b]furan 97%

Lieferant: Apollo Scientific

Synonyme: 5-Chlorobenzofuran, 5-Chloro-1-benzofuran

OR909407-1G OR909407-5G
APOSOR909407-1GEA 364 EUR
APOSOR909407-1G APOSOR909407-5G
5-Chlorobenzo[b]furan 97%
5-Chlorobenzo[b]furan
Formel: C₈H₅ClO
Molecular Weight: 152,578 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699415
CAS-Nummer: 23145-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR