4-Isopropoxypiperidine 96%

Lieferant: Apollo Scientific

OR909415-1G OR909415-250MG
APOSOR909415-1GEA 458 EUR
APOSOR909415-1G APOSOR909415-250MG
4-Isopropoxypiperidine 96%
4-Isopropoxypiperidine
Formel: C₈H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06804555
CAS-Nummer: 43139-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR