1-(2-Methoxyphenyl)piperidine 98%
Lieferant: Apollo Scientific
OR909424-1G
OR909424-25G
OR909424-5G
APOSOR909424-1GEA
109
EUR
APOSOR909424-1G
APOSOR909424-25G
APOSOR909424-5G
1-(2-Methoxyphenyl)piperidine 98%
1-(2-Methoxyphenyl)piperidine
Formel:
C₁₂H₁₇NO Molecular Weight: 191,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20231456 CAS-Nummer: 5181-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...