2-chloro-4-methoxy-1-nitrobenzene 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-4-methoxy-1-nitrobenzene

OR909438-1G OR909438-5G
APOSOR909438-1GEA 81.2 EUR
APOSOR909438-1G APOSOR909438-5G
2-chloro-4-methoxy-1-nitrobenzene 95%
2-chloro-4-methoxy-1-nitrobenzene
Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156594
CAS-Nummer: 28987-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR