1-(tert-Butoxy)-4-nitrobenzene 98%
Lieferant: Apollo Scientific
OR909486-1G
OR909486-25G
OR909486-5G
APOSOR909486-1GEA
132
EUR
APOSOR909486-1G
APOSOR909486-25G
APOSOR909486-5G
1-(tert-Butoxy)-4-nitrobenzene 98%
1-(tert-Butoxy)-4-nitrobenzene
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195.215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094683 CAS-Nummer: 2109-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...