2-Methoxy-4-piperidinopyrimidine 98%

Lieferant: Apollo Scientific

OR909498-10G OR909498-1G OR909498-5G
APOSOR909498-10GEA 754 EUR
APOSOR909498-10G APOSOR909498-1G APOSOR909498-5G
2-Methoxy-4-piperidinopyrimidine 98%
2-Methoxy-4-piperidinopyrimidine
Formel: C₁₀H₁₅N₃O
Molecular Weight: 193,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22375033
CAS-Nummer: 1381944-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR