2-Acetamido-5-methylthiazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(5-Methyl-1,3-thiazol-2-yl)acetamide
OR909503-1G
OR909503-5G
APOSOR909503-1GEA
176
EUR
APOSOR909503-1G
APOSOR909503-5G
2-Acetamido-5-methylthiazole 98%
2-Acetamido-5-methylthiazole
Formel:
C₆H₈N₂OS Molecular Weight: 156,206 g/mol Schmelzpunkt: 230…232 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD18801061 CAS-Nummer: 61996-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...