2-Ethyl-3,4-dihydro-2H-1,4-benzoxazine 98%

Lieferant: Apollo Scientific

OR909508-1G OR909508-250MG OR909508-5G
APOSOR909508-1GEA 328 EUR
APOSOR909508-1G APOSOR909508-250MG APOSOR909508-5G
2-Ethyl-3,4-dihydro-2H-1,4-benzoxazine 98%
2-Ethyl-3,4-dihydro-2H-1,4-benzoxazine
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11603418
CAS-Nummer: 1082930-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR