2-Methoxy-N,N-dimethylpyrimidin-4-amine 98%

Lieferant: Apollo Scientific

OR909509-10G OR909509-1G OR909509-5G
APOSOR909509-10GEA 754 EUR
APOSOR909509-10G APOSOR909509-1G APOSOR909509-5G
2-Methoxy-N,N-dimethylpyrimidin-4-amine 98%
2-Methoxy-N,N-dimethylpyrimidin-4-amine
Formel: C₇H₁₁N₃O
Molecular Weight: 153.182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22375035
CAS-Nummer: 57054-82-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR