3-(2-Oxopyrrolidin-1-yl)benzonitrile 98%

Lieferant: Apollo Scientific

OR909555-10G OR909555-1G OR909555-5G
APOSOR909555-10GEA 754 EUR
APOSOR909555-10G APOSOR909555-1G APOSOR909555-5G
3-(2-Oxopyrrolidin-1-yl)benzonitrile 98%
3-(2-Oxopyrrolidin-1-yl)benzonitrile
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09950471
CAS-Nummer: 939999-23-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR