2-Propanoyl-3-picoline 96%

Lieferant: Apollo Scientific

Synonyme: 1-(3-Methylpyridin-2-yl)propan-1-one

OR909599-1G OR909599-5G
APOSOR909599-1GEA 219 EUR
APOSOR909599-1G APOSOR909599-5G
2-Propanoyl-3-picoline 96%
2-Propanoyl-3-picoline
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16087790
CAS-Nummer: 917872-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR