3-(2,5-Dimethylpyrrol-1-yl)aniline 98%

Lieferant: Apollo Scientific

OR909637-10G OR909637-1G OR909637-5G
APOSOR909637-10GEA 754 EUR
APOSOR909637-10G APOSOR909637-1G APOSOR909637-5G
3-(2,5-Dimethylpyrrol-1-yl)aniline 98%
3-(2,5-Dimethylpyrrol-1-yl)aniline
Formel: C₁₂H₁₄N₂
Molecular Weight: 186,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01125302
CAS-Nummer: 247225-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR