tert-Butyl N-(carbamothioylmethyl)carbamate 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-amino-2-thioxoethylcarbamate, tert-Butyl N-(2-amino-2-sulphanylideneethyl)carbamate, tert-Butyl (2-amino-2-thioxoethyl)carbamate

OR909672-1G OR909672-5G
APOSOR909672-1GEA 75.4 EUR
APOSOR909672-1G APOSOR909672-5G
tert-Butyl N-(carbamothioylmethyl)carbamate 98%
tert-Butyl N-(carbamothioylmethyl)carbamate
Formel: C₇H₁₄N₂O₂S
Molecular Weight: 190,263 g/mol
Schmelzpunkt: 120…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09025922
CAS-Nummer: 89226-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR