Ethyl 3-(pyridin-2-yl)propanoate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-(2-Pyridyl)propanoate

OR909728-1G OR909728-5G
APOSOR909728-1GEA 81.2 EUR
APOSOR909728-1G APOSOR909728-5G
Ethyl 3-(pyridin-2-yl)propanoate 97%
Ethyl 3-(pyridin-2-yl)propanoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04004149
CAS-Nummer: 2739-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR