1-(Methoxymethyl)pyrazole 98%

Lieferant: Apollo Scientific

OR909734-1G OR909734-250MG
APOSOR909734-1GEA 364 EUR
APOSOR909734-1G APOSOR909734-250MG
1-(Methoxymethyl)pyrazole 98%
1-(Methoxymethyl)pyrazole
Formel: C₅H₈N₂O
Molecular Weight: 112,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21195618
CAS-Nummer: 125001-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR