tert-Butyl N-(2-chloroethyl)-N-methylcarbamate 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyl (2-chloroethyl)(methyl)carbamate

OR909760-1G OR909760-5G OR909760-10G
APOSOR909760-1GEA 132 EUR
APOSOR909760-1G APOSOR909760-5G APOSOR909760-10G
tert-Butyl N-(2-chloroethyl)-N-methylcarbamate 96%
tert-Butyl N-(2-chloroethyl)-N-methylcarbamate
Formel: C₈H₁₆ClNO₂
Molecular Weight: 193,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20624610
CAS-Nummer: 220074-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR