6-Bromo-2-propylaminopyridine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR909809-100G
OR909809-25G
OR909809-5G
APOSOR909809-100GEA
905
EUR
APOSOR909809-100G
APOSOR909809-25G
APOSOR909809-5G
6-Bromo-2-propylaminopyridine hydrochloride 97%
6-Bromo-2-propylaminopyridine hydrochloride
Formel:
C₈H₁₁BrN₂·ClH Molecular Weight: 251,551 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756445 CAS-Nummer: 1150271-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...