7-Methoxy-2,3-dihydro-1-benzopyran-4-one 95%
Lieferant: Apollo Scientific
Synonyme:
7-Methoxychroman-4-one
OR909870-1G
OR909870-250MG
APOSOR909870-1GEA
288
EUR
APOSOR909870-1G
APOSOR909870-250MG
7-Methoxy-2,3-dihydro-1-benzopyran-4-one 95%
7-Methoxy-2,3-dihydro-1-benzopyran-4-one
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178,185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00598405 CAS-Nummer: 42327-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...