N-Methyl-2-(2-nitrophenoxy)ethanamine 98%

Lieferant: Apollo Scientific

OR909974-1G OR909974-5G
APOSOR909974-1GEA 168 EUR
APOSOR909974-1G APOSOR909974-5G
N-Methyl-2-(2-nitrophenoxy)ethanamine 98%
N-Methyl-2-(2-nitrophenoxy)ethanamine
Formel: C₉H₁₂N₂O₃
Molecular Weight: 196,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053870
CAS-Nummer: 1038700-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR