1-(N,N-Diethylaminoethyl)pyrazole 96%
Lieferant: Apollo Scientific
OR910113-1G
OR910113-250MG
APOSOR910113-1GEA
364
EUR
APOSOR910113-1G
APOSOR910113-250MG
1-(N,N-Diethylaminoethyl)pyrazole 96%
1-(N,N-Diethylaminoethyl)pyrazole
Formel:
C₉H₁₇N₃ Molecular Weight: 167,251 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12546525 CAS-Nummer: 71033-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...