1-(Thiophen-2-ylmethyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR910124-10G OR910124-1G OR910124-5G
APOSOR910124-10GEA 754 EUR
APOSOR910124-10G APOSOR910124-1G APOSOR910124-5G
1-(Thiophen-2-ylmethyl)pyrrolidine 95%
1-(Thiophen-2-ylmethyl)pyrrolidine
Formel: C₉H₁₃NS
Molecular Weight: 167,271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23699502
CAS-Nummer: 16087-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR