2-Chloro-4-ethylthiazole 95%

Lieferant: Apollo Scientific

OR910238-100MG OR910238-1G OR910238-250MG
APOSOR910238-100MGEA 174 EUR
APOSOR910238-100MG APOSOR910238-1G APOSOR910238-250MG
2-Chloro-4-ethylthiazole 95%
2-Chloro-4-ethylthiazole
Formel: C₅H₆ClNS
Molecular Weight: 147.626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16817473
CAS-Nummer: 98025-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR