2-Bromo-1-methylimidazole-5-carbaldehyde 97%
Lieferant: Apollo Scientific
OR910320-1G
OR910320-5G
APOSOR910320-1GEA
270
EUR
APOSOR910320-1G
APOSOR910320-5G
2-Bromo-1-methylimidazole-5-carbaldehyde 97%
2-Bromo-1-methylimidazole-5-carbaldehyde
Formel:
C₅H₅BrN₂O Molecular Weight: 189,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10696276 CAS-Nummer: 79326-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...