2-(Thiophen-3-yl)acetamide 98%

Lieferant: Apollo Scientific

OR910332-1G OR910332-250MG
APOSOR910332-1GEA 400 EUR
APOSOR910332-1G APOSOR910332-250MG
2-(Thiophen-3-yl)acetamide 98%
2-(Thiophen-3-yl)acetamide
Formel: C₆H₇NOS
Molecular Weight: 141,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00661538
CAS-Nummer: 13781-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR