1-Methyl-4-[(E)-2-nitroethenyl]pyrazole 98%
Lieferant: Apollo Scientific
OR910363-1G
OR910363-5G
APOSOR910363-1GEA
219
EUR
APOSOR910363-1G
APOSOR910363-5G
1-Methyl-4-[(E)-2-nitroethenyl]pyrazole 98%
1-Methyl-4-[(E)-2-nitroethenyl]pyrazole
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,139 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16061742 CAS-Nummer: 3156-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...