tert-Butyl 4-methylpyrazole-1-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-methyl-1H-pyrazole-1-carboxylate

OR910425-1G OR910425-5G
APOSOR910425-1GEA 144 EUR
APOSOR910425-1G APOSOR910425-5G
tert-Butyl 4-methylpyrazole-1-carboxylate 97%
tert-Butyl 4-methylpyrazole-1-carboxylate
Formel: C₉H₁₄N₂O₂
Molecular Weight: 182.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24466467
CAS-Nummer: 121669-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR