6-Methoxy-1,2-benzoxazol-3-amine 98%

Lieferant: Apollo Scientific

Synonyme: 6-Methoxybenzo[d]isoxazol-3-amine

OR910452-1G OR910452-250MG
APOSOR910452-1GEA 230 EUR
APOSOR910452-1G APOSOR910452-250MG
6-Methoxy-1,2-benzoxazol-3-amine 98%
6-Methoxy-1,2-benzoxazol-3-amine
Formel: C₈H₈N₂O₂
Molecular Weight: 164,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18801052
CAS-Nummer: 157368-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR