3-Formyl-1H-indole-4-carboxylic acid 96%

Lieferant: Apollo Scientific

OR910511-1G OR910511-250MG
APOSOR910511-1GEA 301 EUR
APOSOR910511-1G APOSOR910511-250MG
3-Formyl-1H-indole-4-carboxylic acid 96%
3-Formyl-1H-indole-4-carboxylic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189.167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864030
CAS-Nummer: 208772-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR