1H-1,2,3-Benzotriazole-5-carbonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
1H-Benzo[d][1,2,3]triazole-5-carbonitrile
OR910546-1G
OR910546-5G
APOSOR910546-1GEA
60.9
EUR
APOSOR910546-1G
APOSOR910546-5G
1H-1,2,3-Benzotriazole-5-carbonitrile 98%
1H-1,2,3-Benzotriazole-5-carbonitrile
Formel:
C₇H₄N₄ Molecular Weight: 144,133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669484 CAS-Nummer: 24611-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...