1-Methyl-1,2,3-benzotriazole-5-carbonitrile 96%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-1H-benzo[d][1,2,3]triazole-5-carbonitrile

OR910554-1G OR910554-5G
APOSOR910554-1GEA 82.8 EUR
APOSOR910554-1G APOSOR910554-5G
1-Methyl-1,2,3-benzotriazole-5-carbonitrile 96%
1-Methyl-1,2,3-benzotriazole-5-carbonitrile
Formel: C₈H₆N₄
Molecular Weight: 158,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054186
CAS-Nummer: 1065100-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR