1-Propyl-1,3-benzodiazole-5-carbonitrile 98%

Lieferant: Apollo Scientific

OR910557-1G OR910557-25G OR910557-5G
APOSOR910557-1GEA 145 EUR
APOSOR910557-1G APOSOR910557-25G APOSOR910557-5G
1-Propyl-1,3-benzodiazole-5-carbonitrile 98%
1-Propyl-1,3-benzodiazole-5-carbonitrile
Formel: C₁₁H₁₁N₃
Molecular Weight: 185,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21123473
CAS-Nummer: 1403483-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR