1-Propyl-1,2,3-benzotriazole-5-carbonitrile 98%

Lieferant: Apollo Scientific

OR910559-1G OR910559-25G OR910559-5G
APOSOR910559-1GEA 145 EUR
APOSOR910559-1G APOSOR910559-25G APOSOR910559-5G
1-Propyl-1,2,3-benzotriazole-5-carbonitrile 98%
1-Propyl-1,2,3-benzotriazole-5-carbonitrile
Formel: C₁₀H₁₀N₄
Molecular Weight: 186,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22421517
CAS-Nummer: 1400644-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR