1-Isopropyl-1,2,3-benzotriazole-5-carbonitrile 98%

Lieferant: Apollo Scientific

OR910561-10G OR910561-1G OR910561-5G
APOSOR910561-10GEA 377 EUR
APOSOR910561-10G APOSOR910561-1G APOSOR910561-5G
1-Isopropyl-1,2,3-benzotriazole-5-carbonitrile 98%
1-Isopropyl-1,2,3-benzotriazole-5-carbonitrile
Formel: C₁₀H₁₀N₄
Molecular Weight: 186,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09031714
CAS-Nummer: 879558-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR