1-Ethyl-5-nitroindazole 98%
Lieferant: Apollo Scientific
Synonyme:
1-Ethyl-5-nitro-1H-indazole
OR910594-1G
APOSOR910594-1GEA
119
EUR
APOSOR910594-1G
1-Ethyl-5-nitroindazole 98%
1-Ethyl-5-nitroindazole
Formel:
C₉H₉N₃O₂ Molecular Weight: 191,187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20491412 CAS-Nummer: 5228-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...