5-Cyanogramine 98%

Lieferant: Apollo Scientific

OR910601-1G OR910601-5G
APOSOR910601-1GEA 248 EUR
APOSOR910601-1G APOSOR910601-5G
5-Cyanogramine 98%
5-Cyanogramine
Formel: C₁₂H₁₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01719098
CAS-Nummer: 25514-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR