1,2,3-Benzotriazol-2-ylacetic acid 95%

Lieferant: Apollo Scientific

OR910623-1G OR910623-5G
APOSOR910623-1GEA 248 EUR
APOSOR910623-1G APOSOR910623-5G
1,2,3-Benzotriazol-2-ylacetic acid 95%
1,2,3-Benzotriazol-2-ylacetic acid
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01812492
CAS-Nummer: 4144-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR