4-(3,5-Dimethylphenyl)aniline 98%

Lieferant: Apollo Scientific

OR910648-1G OR910648-250MG
APOSOR910648-1GEA 632 EUR
APOSOR910648-1G APOSOR910648-250MG
4-(3,5-Dimethylphenyl)aniline 98%
4-(3,5-Dimethylphenyl)aniline
Formel: C₁₄H₁₅N
Molecular Weight: 197,276 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03792760
CAS-Nummer: 444143-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR