4-(2-Methylphenyl)benzonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 2'-Methyl-[1,1'-biphenyl]-4-carbonitrile

OR910849-1G OR910849-5G
APOSOR910849-1GEA 219 EUR
APOSOR910849-1G APOSOR910849-5G
4-(2-Methylphenyl)benzonitrile 98%
4-(2-Methylphenyl)benzonitrile
Formel: C₁₄H₁₁N
Molecular Weight: 193.244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05981801
CAS-Nummer: 189828-30-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR