5-(2-Methylphenyl)pyridin-2-amine 98%

Lieferant: Apollo Scientific

OR910865-1G OR910865-5G
APOSOR910865-1GEA 219 EUR
APOSOR910865-1G APOSOR910865-5G
5-(2-Methylphenyl)pyridin-2-amine 98%
5-(2-Methylphenyl)pyridin-2-amine
Formel: C₁₂H₁₂N₂
Molecular Weight: 184,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06802147
CAS-Nummer: 503536-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR