[4-(4-Methoxyphenyl)phenyl]acetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(4'-Methoxy-[1,1'-biphenyl]-4-yl)acetic acid

OR910887-1G OR910887-5G
APOSOR910887-1GEA 208 EUR
APOSOR910887-1G APOSOR910887-5G
[4-(4-Methoxyphenyl)phenyl]acetic acid 98%
[4-(4-Methoxyphenyl)phenyl]acetic acid
Formel: C₁₅H₁₄O₃
Molecular Weight: 242.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426471
CAS-Nummer: 60277-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR