2-N-Butylamino-5-nitropyridine 98%

Lieferant: Apollo Scientific

OR911056-1G OR911056-25G OR911056-5G
APOSOR911056-1GEA 145 EUR
APOSOR911056-1G APOSOR911056-25G APOSOR911056-5G
2-N-Butylamino-5-nitropyridine 98%
2-N-Butylamino-5-nitropyridine
Formel: C₉H₁₃N₃O₂
Molecular Weight: 195.218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11116745
CAS-Nummer: 26820-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR