2-Methanesulphonylpyridine-4-carbonitrile 96%

Lieferant: Apollo Scientific

OR911079-1G OR911079-250MG
APOSOR911079-1GEA 435 EUR
APOSOR911079-1G APOSOR911079-250MG
2-Methanesulphonylpyridine-4-carbonitrile 96%
2-Methanesulphonylpyridine-4-carbonitrile
Formel: C₇H₆N₂O₂S
Molecular Weight: 182.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14705772
CAS-Nummer: 66154-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR