1-Cyclopentyl-4-nitro-1H-pyrazole 98%

Lieferant: Apollo Scientific

OR911107-10G OR911107-1G OR911107-25G
APOSOR911107-10GEA 377 EUR
APOSOR911107-10G APOSOR911107-1G APOSOR911107-25G
1-Cyclopentyl-4-nitro-1H-pyrazole 98%
1-Cyclopentyl-4-nitro-1H-pyrazole
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16345430
CAS-Nummer: 1245772-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR