4-Chloro-2-[(1E)-(hydroxyimino)methyl]phenol 98%

Lieferant: Apollo Scientific

OR911128-1G OR911128-5G
APOSOR911128-1GEA 219 EUR
APOSOR911128-1G APOSOR911128-5G
4-Chloro-2-[(1E)-(hydroxyimino)methyl]phenol 98%
4-Chloro-2-[(1E)-(hydroxyimino)methyl]phenol
Formel: C₇H₆ClNO₂
Molecular Weight: 171.581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412413
CAS-Nummer: 87974-49-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR