(3-Ethoxyphenyl)acetic acid 98%

Lieferant: Apollo Scientific

OR911178-1G OR911178-250MG
APOSOR911178-1GEA 328 EUR
APOSOR911178-1G APOSOR911178-250MG
(3-Ethoxyphenyl)acetic acid 98%
(3-Ethoxyphenyl)acetic acid
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075781
CAS-Nummer: 72775-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR