N-Cyclopropylthiophene-3-carboxamide 98%

Lieferant: Apollo Scientific

OR911185-1G OR911185-5G
APOSOR911185-1GEA 270 EUR
APOSOR911185-1G APOSOR911185-5G
N-Cyclopropylthiophene-3-carboxamide 98%
N-Cyclopropylthiophene-3-carboxamide
Formel: C₈H₉NOS
Molecular Weight: 167,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10674150
CAS-Nummer: 1090897-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR