1-[(Thiophen-3-yl)carbonyl]piperidine 97%

Lieferant: Apollo Scientific

OR911201-1G OR911201-5G
APOSOR911201-1GEA 219 EUR
APOSOR911201-1G APOSOR911201-5G
1-[(Thiophen-3-yl)carbonyl]piperidine 97%
1-[(Thiophen-3-yl)carbonyl]piperidine
Formel: C₁₀H₁₃NOS
Molecular Weight: 195,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20355792
CAS-Nummer: 862463-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR