[(2-Methoxyphenyl)amino]acetic acid 98%
Lieferant: Apollo Scientific
OR911243-1G
OR911243-5G
APOSOR911243-1GEA
255
EUR
APOSOR911243-1G
APOSOR911243-5G
[(2-Methoxyphenyl)amino]acetic acid 98%
[(2-Methoxyphenyl)amino]acetic acid
Formel:
C₉H₁₁NO₃ Molecular Weight: 181.189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00504580 CAS-Nummer: 94800-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...